2,2,2-Trifluoro-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone 97%

Supplier: Ambeed
Warning

A418525-250MG A418525-1G A418525-5G
103827-358EA 100 USD
103827-358 103838-402 76829-428 76842-008 76842-010
2,2,2-Trifluoro-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone 97%
2,2,2-Trifluoro-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₄H₁₆BF₃O₃
MW: 300.08 g/mol
Boiling Pt: 349.5±42.0 °C (760 mm Hg)
Flash Pt: 165.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18915048
CAS Number: 631909-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR