2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(Trichloroacetyl)pyrrole, 2-(trichloroacetyl)-1h-pyrrole
A170307-1000G
A170307-100G
A170307-10G
A170307-500G
A170307-25G
A170307-5G
103809-024EA
439
USD
103809-024
103844-740
77081-850
77247-968
77247-970
77247-972
77301-034
77301-036
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone 98%
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₆H₄Cl₃NO MW: 212.46 g/mol Boiling Pt: 285.3 °C (760 mm Hg) Melting Pt: 72…74 °C Flash Pt: 126.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00128757 CAS Number: 35302-72-8 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...