1,4-Bis(2-hydroxyethyl)piperazine 97%

Supplier: Ambeed
Warning

Synonyms: 2,2'-(Piperazine-1,4-diyl)diethanol

A119679-100G A119679-10G A119679-25G A119679-500G A119679-5G
103808-210EA 93 USD
103808-210 77189-606 77189-608 77189-610 77189-612 77297-714
1,4-Bis(2-hydroxyethyl)piperazine 97%
1,4-Bis(2-hydroxyethyl)piperazine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MW: 174.24 g/mol
Boiling Pt: 305.7 °C (760 mm Hg)
Melting Pt: 133.5…136.0 °C
Flash Pt: 166.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00006157
CAS Number: 122-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR