2,2'-(Piperazine-1,4-diyl)diethanamine 95%
Supplier: Ambeed
|
Danger
|
A386260-1G
A386260-100MG
A386260-250MG
A386260-5G
103826-794EA
192
USD
103826-794
76841-186
77264-310
77264-312
77264-314
2,2'-(Piperazine-1,4-diyl)diethanamine 95%
2,2'-(Piperazine-1,4-diyl)diethanamine
Precaución: for Research Use Only
Formula:
C₈H₂₀N₄ MW: 172.27 g/mol Boiling Pt: 302.2 °C (760 mm Hg) Flash Pt: 151.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00720062 CAS Number: 6531-38-0 UN: 1759 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...