2,2'-(Octylazanediyl)diethanol 95%

Supplier: Ambeed
Warning

A789263-1G A789263-250MG
103841-010EA 77 USD
103841-010 76845-264 77255-852
2,2'-(Octylazanediyl)diethanol 95%
2,2'-(Octylazanediyl)diethanol

Precaución: for Research Use Only

Formula: C₁₂H₂₇NO₂
MW: 217.35 g/mol
Boiling Pt: 342.4 °C (760 mm Hg)
Flash Pt: 150.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06252160
CAS Number: 15520-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR