Stearyldiethanolamine 97%

Supplier: Ambeed
Warning

Synonyms: 2,2'-(Octadecylimino)diethanol, 2,2'-(Octadecylazanediyl)diethanol

A204410-10G A204410-25G A204410-100G A204410-500G
103848-214EA 6 USD
103848-214 76785-450 77086-388 77567-960 77667-712
Stearyldiethanolamine 97%
Stearyldiethanolamine

Precaución: For research use only.

Formula: C₂₂H₄₇NO₂
MW: 357.62 g/mol
Boiling Pt: 452.1 °C (760 mm Hg)
Melting Pt: 51 °C
Flash Pt: 174.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00059003
CAS Number: 10213-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR