N-Methyliminodiacetic acid 98%

Supplier: Ambeed
Warning

A101828-100G A101828-10G A101828-25G A101828-500G A101828-5G A101828-1000G
103821-852EA 5 USD
103821-852 76717-580 76717-582 76717-584 76717-586 77282-840 77599-488
N-Methyliminodiacetic acid 98%
N-Methyliminodiacetic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₄
Boiling Pt: 332.8 °C (760 mm Hg)
Melting Pt: 220 °C
Flash Pt: 155.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00004284
CAS Number: 4408-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR