2,2'-Ethylenedianiline 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-(Ethane-1,2-diyl)dianiline, alpha,alpha'-Bi-o-toluidine, 2,2'-Diamino-1,2-diphenylethane, 2,2'-Diaminobibenzyl
A263069-1G
A263069-25G
A263069-5G
A263069-250MG
A263069-100G
103812-602EA
15
USD
103812-602
103852-498
77092-124
77092-126
77092-128
77193-882
77746-445
2,2'-Ethylenedianiline 95%
2,2'-Ethylenedianiline
Precaución: for Research Use Only
Formula:
C₁₄H₁₆N₂ MW: 212.30 g/mol Boiling Pt: 382.4 °C (760 mm Hg) Melting Pt: 72…77 °C Flash Pt: 221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00127990 CAS Number: 34124-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...