2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 9,9-Dioctylfluorene-2,7-diboronic acid bis(pinacol) ester
A230133-100G
A230133-1G
A230133-25G
A230133-5G
103819-524EA
8
USD
103819-524
103850-424
76987-312
77089-276
77089-278
77089-280
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene 97%
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene
Precaución: for Research Use Only
Formula:
C₄₁H₆₄B₂O₄ Boiling Pt: 701.6 °C (760 mm Hg) Flash Pt: 378.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD16294554 CAS Number: 196207-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...