3,5-Bis(2-cyanoprop-2-yl)toluene 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-(5-Methyl-1,3-phenylene)bis(2-methylpropanenitrile), Anastrozole EP Impurity H

A203204-100G A203204-10G A203204-1G A203204-25G A203204-5G
103817-444EA 6 USD
103817-444 103848-098 76785-420 76785-422 77086-216 77086-218 77086-220
3,5-Bis(2-cyanoprop-2-yl)toluene 98%
3,5-Bis(2-cyanoprop-2-yl)toluene

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂
MW: 226.317 g/mol
Boiling Pt: 339.1 °C (760 mm Hg)
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD08062446
CAS Number: 120511-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR