1,3-Phenylenediacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,2'-(1,3-Phenylene)diacetic acid

A221806-250MG A221806-1G A221806-25G A221806-5G
103818-946EA 6 USD
103818-946 103849-826 77192-908 77303-612 77303-614 77303-616
1,3-Phenylenediacetic acid 97%
1,3-Phenylenediacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 401.6 °C (760 mm Hg)
Melting Pt: 175…177 °C
Flash Pt: 215.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00004341
CAS Number: 19806-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR