N,N'-Bis(salicylidene)ethylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: Ethylenebis(salicylimine), 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))diphenol, SALEN

A260347-10G A260347-25G A260347-5G
103852-374EA 7 USD
103852-374 77031-750 77091-958 77091-960
N,N'-Bis(salicylidene)ethylenediamine 98%
N,N'-Bis(salicylidene)ethylenediamine

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₂
MW: 268.31 g/mol
Melting Pt: 127…128 °C
Flash Pt: 193.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002244
CAS Number: 94-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR