(Triphenylphosphoranylidene)acetaldehyde 97%

Supplier: Ambeed
Warning

A150408-10G A150408-500G A150408-5G A150408-25G A150408-100G A150408-1000G
103842-298EA 11 USD
103842-298 76689-340 76689-342 77299-804 77317-812 77549-888 77877-012
(Triphenylphosphoranylidene)acetaldehyde 97%
(Triphenylphosphoranylidene)acetaldehyde

Precaución: for Research Use Only

Formula: C₂₀H₁₇OP
Boiling Pt: 472.6 °C (760 mm Hg)
Melting Pt: 185…188 °C
Flash Pt: 239.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00006994
CAS Number: 2136-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR