2-(Trimethylsilyl)thiazole 95% (by GC)

Supplier: Ambeed
Warning

A131155-10G A131155-25G A131155-5G
103801-534EA 24 USD
103801-534 77136-920 77298-450 77298-452
2-(Trimethylsilyl)thiazole 95% (by GC)
2-(Trimethylsilyl)thiazole

Precaución: for Research Use Only

Formula: C₆H₁₁NSSi
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066274
CAS Number: 79265-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR