2-(Trimethylsilyl)phenyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

Synonyms: Trifluoromethanesulphonic acid 2-(Trimethylsilyl)phenyl Ester, 2-(Trimethylsilyl)phenyl Triflate

A294211-1G A294211-25G A294211-5G A294211-10G A294211-250MG
103814-004EA 19 USD
103814-004 103835-144 76837-776 76837-778 76837-780 77250-734 77250-736
2-(Trimethylsilyl)phenyl trifluoromethanesulfonate 95%
2-(Trimethylsilyl)phenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₁₃F₃O₃SSi
MW: 298.35 g/mol
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799598
CAS Number: 88284-48-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR