2-(Trifluoromethoxy)phenol 97%
Supplier: Ambeed
|
Warning
|
A111982-10G
A111982-25G
A111982-5G
103823-758EA
10
USD
103823-758
76995-264
76995-266
77310-566
2-(Trifluoromethoxy)phenol 97%
2-(Trifluoromethoxy)phenol
Precaución: for Research Use Only
Formula:
C₇H₅F₃O₂ Boiling Pt: 156.8 °C (760 mm Hg) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236324 CAS Number: 32858-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...