2-(Trifluoromethyl)thiazole-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A124263-250MG A124263-1G
103800-510EA 221 USD
103800-510 77189-824 77297-980
2-(Trifluoromethyl)thiazole-5-carbaldehyde 95%
2-(Trifluoromethyl)thiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₂F₃NOS
MW: 181.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08448192
CAS Number: 903130-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR