2-(Trifluoromethyl)terephthalic acid 95%

Supplier: Ambeed
Warning

A371202-100MG A371202-250MG A371202-1G
103826-478EA 168 USD
103826-478 103837-498 76989-890 76989-892 77306-830
2-(Trifluoromethyl)terephthalic acid 95%
2-(Trifluoromethyl)terephthalic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD06208277
CAS Number: 1483-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR