2-(Trifluoromethyl)pyrimidine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A273033-25G A273033-5G A273033-10G A273033-250MG
103813-120EA 40 USD
103813-120 103853-036 77092-828 77092-830 77250-386 77250-388
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid 97%
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃F₃N₂O₂
Boiling Pt: 220.6 °C (760 mm Hg)
Melting Pt: 170…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09054851
CAS Number: 306960-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR