2-(Trifluoromethyl)pyrimidine-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A217928-250MG A217928-1G A217928-25G A217928-5G
103818-670EA 64 USD
103818-670 103849-402 76785-874 77088-046 77088-048 77088-050
2-(Trifluoromethyl)pyrimidine-5-carbaldehyde 97%
2-(Trifluoromethyl)pyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃F₃N₂O
MW: 176.09 g/mol
Boiling Pt: 153.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08752498
CAS Number: 304693-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR