2-(Trifluoromethyl)pyrimidin-5-ol 97%
Supplier: Ambeed
|
Warning
|
A126636-100MG
A126636-250MG
A126636-1G
103800-846EA
106
USD
103800-846
77271-814
77271-816
77313-076
2-(Trifluoromethyl)pyrimidin-5-ol 97%
2-(Trifluoromethyl)pyrimidin-5-ol
Precaución: for Research Use Only
Formula:
C₅H₃F₃N₂O MW: 164.085 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD10696944 CAS Number: 100991-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...