2-(Trifluoromethyl)pyrimidin-5-amine 98%

Supplier: Ambeed
Warning

A339315-1G A339315-10G A339315-250MG
103825-788EA 45 USD
103825-788 103836-764 76839-784 77251-372 77251-374
2-(Trifluoromethyl)pyrimidin-5-amine 98%
2-(Trifluoromethyl)pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₅H₄F₃N₃
MW: 163.1000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09754047
CAS Number: 73418-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR