2-(Trifluoromethyl)pyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A119928-100MG A119928-250MG A119928-1G
103799-864EA 187 USD
103799-864 77026-322 77026-324 77311-972
2-(Trifluoromethyl)pyrimidin-4-amine 95%
2-(Trifluoromethyl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₄F₃N₃
MW: 163.101 g/mol
Boiling Pt: 155.3 °C (760 mm Hg)
Melting Pt: 180…181 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436606
CAS Number: 672-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR