2-(Trifluoromethyl)pyridine-5-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 6-(Trifluoromethyl)nicotinaldehyde

A364922-25G A364922-5G A364922-100MG A364922-10G
103826-372EA 15 USD
103826-372 103837-392 76840-624 76840-626 77167-894 77167-896
2-(Trifluoromethyl)pyridine-5-carboxaldehyde 98%
2-(Trifluoromethyl)pyridine-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₃NO
MW: 175.11 g/mol
Boiling Pt: 205.3 °C (760 mm Hg)
Melting Pt: 52…56 °C
Flash Pt: 93.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862647
CAS Number: 386704-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR