2-(Trifluoromethyl)pyridin-4-ol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Hydroxy-2-(trifluoromethyl)pyridine
A203799-100G
A203799-10G
A203799-250MG
A203799-25G
A203799-1G
A203799-5G
103817-494EA
7
USD
103817-494
103848-152
77030-214
77030-216
77030-218
77030-220
77086-288
77086-290
2-(Trifluoromethyl)pyridin-4-ol 97%
2-(Trifluoromethyl)pyridin-4-ol
Precaución: for Research Use Only
Formula:
C₆H₄F₃NO MW: 163.10 g/mol Boiling Pt: 324.4 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD01571202 CAS Number: 170886-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...