3-Amino-2-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Trifluoromethyl)-3-pyridylamine, 2-(Trifluoromethyl)pyridin-3-amine

A127153-1G A127153-5G A127153-250MG
103800-922EA 168 USD
103800-922 77298-182 77298-184 77614-992
3-Amino-2-(trifluoromethyl)pyridine 97%
3-Amino-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₅F₃N₂
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD09753488
CAS Number: 106877-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR