2-(Trifluoromethyl)phenylhydrazinium chloride 98%

Warning

Synonyms: 2-(Trifluoromethyl)phenylhydrazine hydrochloride, 1-(2-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 2-Hydrazinobenzotrifluoride hydrochloride, (2-(Trifluormethyl)phenyl)hydrazin HCl

A220238-10G A220238-5G A220238-25G
103818-846 103849-728 76770-792 77088-316 77303-544
2-(Trifluoromethyl)phenylhydrazinium chloride 98%
2-(Trifluoromethyl)phenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₇H₈ClF₃N₂
MW: 212.6 g/mol
Melting Pt: 220 °C
Flash Pt: 70.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00102619
CAS Number: 3107-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR