(α,α,α-Trifluoro-2-tolyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Trifluoromethyl)phenylacetic acid, (2-(Trifluoromethyl)phenyl)acetic acid

A100013-100G A100013-10G A100013-1G A100013-25G A100013-5G
103820-918EA 11 USD
103820-918 76686-966 76686-968 76686-970 76686-972 77296-534
(α,α,α-Trifluoro-2-tolyl)acetic acid 97%
(α,α,α-Trifluoro-2-tolyl)acetic acid

Precaución: for Research Use Only

Formula: F₃CC₆H₄CH₂COOH
MW: 204.15 g/mol
Boiling Pt: 249.9 °C (760 mm Hg)
Melting Pt: 100…102 °C
Flash Pt: 73.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004327
CAS Number: 3038-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR