2-(Trifluoromethyl)isonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Trifluoromethyl)pyridine-4-carboxylic acid

A117951-100G A117951-10G A117951-250MG A117951-25G A117951-5G A117951-1G
103798-960EA 10 USD
103798-960 103824-850 77245-986 77245-988 77245-990 77245-992 77297-564 77311-632
2-(Trifluoromethyl)isonicotinic acid 98%
2-(Trifluoromethyl)isonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Boiling Pt: 338.8 °C (760 mm Hg)
Melting Pt: 217…223 °C
Flash Pt: 112.8 °C
Storage Temperature: Ambient
MDL Number: MFCD07774132
CAS Number: 131747-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR