α,α,α-Trifluoro-o-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Trifluoromethyl)benzoic acid

A145347-100G A145347-10G A145347-500G A145347-5G A145347-25G A145347-1000G
103841-608EA 5 USD
103841-608 77235-166 77235-168 77235-170 77299-448 77316-890 77549-764
α,α,α-Trifluoro-o-toluic acid 97%
α,α,α-Trifluoro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
Boiling Pt: 243.7 °C (760 mm Hg)
Melting Pt: 107…110 °C
Flash Pt: 101.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002476
CAS Number: 433-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR