2-(Trifluoromethyl)benzamide 98%
Supplier: Ambeed
|
Warning
|
A128037-100G
A128037-10G
A128037-5G
A128037-1G
A128037-25G
103801-058EA
9
USD
103801-058
77234-372
77234-374
77298-260
77313-344
77313-346
2-(Trifluoromethyl)benzamide 98%
2-(Trifluoromethyl)benzamide
Precaución: for Research Use Only
Formula:
C₈H₆F₃NO Boiling Pt: 376 °C (760 mm Hg) Melting Pt: 160…161 °C Flash Pt: 181.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014798 CAS Number: 360-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...