α,α,α-Trifluoro-2-tolualdehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Trifluoromethyl)benzaldehyde
A329711-25G
A329711-5G
A329711-1000G
A329711-100G
A329711-10G
A329711-500G
103836-494EA
5
USD
103836-494
76839-466
76839-468
76978-212
76978-214
76978-216
76978-218
α,α,α-Trifluoro-2-tolualdehyde 98%
α,α,α-Trifluoro-2-tolualdehyde
Precaución: For research use only
Formula:
C₈H₅F₃O MW: 174.12 g/mol Boiling Pt: 194.3 °C (760 mm Hg) Melting Pt: 40 °C Flash Pt: 61.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003337 CAS Number: 447-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...