2-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[1,5-a]pyrazine 95%
Supplier: Ambeed
|
Warning
|
A179532-100MG
A179532-250MG
A179532-1G
103807-440EA
295
USD
103807-440
103845-790
77248-218
77248-220
77301-618
2-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[1,5-a]pyrazine 95%
2-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[1,5-a]pyrazine
Precaución: for Research Use Only
Formula:
C₆H₇F₃N₄ MW: 192.1400 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09834979 CAS Number: 681249-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...