2-(Trifluoromethyl)benzimidazole 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Trifluoromethyl)-1H-benzimidazole, 2-(Trifluoromethyl)-1H-benzo[d]imidazole, TFB-2

A238356-1G A238356-10G A238356-250MG
103811-222EA 9 USD
103811-222 103851-002 77090-116 77238-046 77238-048
2-(Trifluoromethyl)benzimidazole 95%
2-(Trifluoromethyl)benzimidazole

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
Boiling Pt: 262.8 °C (760 mm Hg)
Melting Pt: 208…211 °C
Flash Pt: 155.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00022675
CAS Number: 312-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR