2-(Trifluoromethoxy)benzonitrile 98%

Supplier: Ambeed
Danger

A146580-10G A146580-25G A146580-5G
103841-776EA 10 USD
103841-776 77110-068 77317-108 77317-110
2-(Trifluoromethoxy)benzonitrile 98%
2-(Trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Boiling Pt: 201.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00042409
CAS Number: 63968-85-4
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR