2-(Trifluoromethoxy)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Trifluoromethoxy)benzene-1-sulphonyl chloride

A823479-25G A823479-5G A823479-10G
103841-146EA 10 USD
103841-146 76845-384 76845-386 77036-554
2-(Trifluoromethoxy)benzenesulfonyl chloride 98%
2-(Trifluoromethoxy)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 294.8 °C (760 mm Hg)
Melting Pt: 27…31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052316
CAS Number: 103008-51-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR