2-(Trifluoromethoxy)benzaldehyde 98%

Supplier: Ambeed
Warning

A199511-100G A199511-10G A199511-25G
103847-734EA 11 USD
103847-734 76770-096 76770-098 77302-620
2-(Trifluoromethoxy)benzaldehyde 98%
2-(Trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
Flash Pt: 67.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00042405
CAS Number: 94651-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR