trans-4-(Boc-amino)cyclohexaneacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(trans-4-((tert-Butoxycarbonyl)amino)cyclohexyl)acetic acid, Boc-1,4-trans-acha-oh

A184699-100MG A184699-10G A184699-250MG A184699-1G A184699-25G A184699-5G
103815-772EA 12 USD
103815-772 103846-336 76690-670 76690-672 76690-674 77083-950 77083-952 77083-954
trans-4-(Boc-amino)cyclohexaneacetic acid 97%
trans-4-(Boc-amino)cyclohexaneacetic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Boiling Pt: 414.9 °C (760 mm Hg)
Flash Pt: 204.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03844588
CAS Number: 189153-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR