Thiophen-3-acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Thiophen-3-yl)acetonitrile, 3-Thiopheneacetonitrile

A199268-100G A199268-1G A199268-25G A199268-5G
103817-088EA 16 USD
103817-088 103847-712 76999-266 77085-752 77085-754 77085-756
Thiophen-3-acetonitrile 98%
Thiophen-3-acetonitrile

Precaución: for Research Use Only

Formula: C₆H₅NS
MW: 123.18 g/mol
Boiling Pt: 124…125 °C (760 mm Hg)
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00005471
CAS Number: 13781-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR