2-(2-Thienyl)ethyl-p-toluenesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Thiophen-2-yl)ethyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid 2-(2-Thienyl)ethyl Ester

A225237-100G A225237-10G A225237-5G A225237-25G
103809-592EA 250 USD
103809-592 77030-922 77030-924 77030-926 77088-744
2-(2-Thienyl)ethyl-p-toluenesulfonate 97%
2-(2-Thienyl)ethyl-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₃S₂
MW: 282.37 g/mol
Boiling Pt: 433.2 °C (760 mm Hg)
Flash Pt: 215.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05150366
CAS Number: 40412-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR