Thiophene-2-ethylamine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(Thiophen-2-yl)ethanamine, 2-(2-Aminoethyl)thiophene
A257898-25G
A257898-1000G
A257898-100G
A257898-10G
A257898-500G
A257898-5G
103852-216EA
5
USD
103852-216
77091-738
77113-594
77113-596
77113-598
77113-600
77304-722
Thiophene-2-ethylamine 98%
Thiophene-2-ethylamine
Precaución: for Research Use Only
Formula:
C₆H₉NS MW: 127.21 g/mol Boiling Pt: 199.7 °C (760 mm Hg) Flash Pt: 74.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051495 CAS Number: 30433-91-1 UN: 2735 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...