2-Thienylacetic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-Thiopheneacetic acid, Thiophene-2-acetic acid, 2-(Thiophen-2-yl)acetic acid

A316804-100MG A316804-5G A316804-25G
103836-050EA 8 USD
103836-050 77290-240 77290-242 77306-154
2-Thienylacetic acid 98%
2-Thienylacetic acid

Precaución: for Research Use Only

Formula: C₆H₆O₂S
MW: 142.18 g/mol
Boiling Pt: 269.2 °C (760 mm Hg)
Melting Pt: 63…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005458
CAS Number: 1918-77-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR