2-(Tetrahydro-2H-pyran-4-yloxy)pyridine-5-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)pyridine

A136960-100MG A136960-250MG
103802-384EA 75 USD
103802-384 77246-738 77246-740
2-(Tetrahydro-2H-pyran-4-yloxy)pyridine-5-boronic acid pinacol ester 95%
2-(Tetrahydro-2H-pyran-4-yloxy)pyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₄
MW: 305.18 g/mol
Boiling Pt: 419.9 °C (760 mm Hg)
Flash Pt: 207.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09702390
CAS Number: 910036-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR