2-(tert-Butyl)isonicotinic acid 96%

Supplier: Ambeed
Warning

A483379-100MG A483379-1G A483379-5G
103828-156EA 99 USD
103828-156 103839-230 76818-630 77307-550 77307-552
2-(tert-Butyl)isonicotinic acid 96%
2-(tert-Butyl)isonicotinic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
Boiling Pt: 355.5±30.0 °C (760 mm Hg)
Flash Pt: 168.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04113815
CAS Number: 91940-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR