2-tert-Butylaniline 98%
Supplier: Ambeed
|
Warning
|
A271659-25G
A271659-5G
A271659-100G
A271659-10G
A271659-1G
A271659-500G
103813-054EA
12
USD
103813-054
103852-966
77092-746
77092-748
77276-784
77276-786
77305-086
77571-676
2-tert-Butylaniline 98%
2-tert-Butylaniline
Precaución: for Research Use Only
Formula:
C₁₀H₁₅N Boiling Pt: 123…124 °C (760 mm Hg) Melting Pt: …60 °C Flash Pt: 102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130023 CAS Number: 6310-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...