tert-Butylboronic acid, pinacol ester 97%

Supplier: Ambeed
Danger

Synonyms: 2-(tert-Butyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A118982-10G A118982-1G A118982-5G A118982-25G
103799-148EA 14 USD
103799-148 76822-296 77311-818 77311-820 77571-120
tert-Butylboronic acid, pinacol ester 97%
tert-Butylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₂₁BO₂
MW: 184.084 g/mol
Boiling Pt: 186.5 °C (760 mm Hg)
Flash Pt: 66.604 °C
Storage Temperature: Freezer
MDL Number: MFCD06657801
CAS Number: 99810-76-1
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR