2-tert-Butyl-1,1,3,3-tetramethylguanidine 96%

Supplier: Ambeed
Danger

Synonyms: 2-(tert-Butyl)-1,1,3,3-tetramethylguanidine, BTMG

A165559-10G A165559-1G A165559-5G
103805-974EA 5 USD
103805-974 103844-228 77163-338 77300-790 77320-442
2-tert-Butyl-1,1,3,3-tetramethylguanidine 96%
2-tert-Butyl-1,1,3,3-tetramethylguanidine

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₉H₂₁N₃
MW: 171.29 g/mol
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862972
CAS Number: 29166-72-1
UN: 3267
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR