tert-Butoxy acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(tert-Butoxy)acetic acid

A121695-100G A121695-10G A121695-25G A121695-1G A121695-5G
103800-108EA 20 USD
103800-108 77149-216 77149-218 77149-220 77312-232 77312-234
tert-Butoxy acetic acid 95%
tert-Butoxy acetic acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 222.3 °C (760 mm Hg)
Flash Pt: 88.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03452738
CAS Number: 13211-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR