2-(Quinolin-6-yl)acetic acid 97%

Supplier: Ambeed
Danger

A100734-10G A100734-250MG A100734-25G A100734-1G A100734-5G
103795-600EA 17 USD
103795-600 103821-296 76687-008 76687-010 76687-012 77282-552 77282-554
2-(Quinolin-6-yl)acetic acid 97%
2-(Quinolin-6-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
Boiling Pt: 388.7 °C (760 mm Hg)
Flash Pt: 188.9 °C
Storage Temperature: Ambient
MDL Number: MFCD07776794
CAS Number: 5622-34-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR