2-(Pyrimidin-5-yl)benzaldehyde 95%

Supplier: Ambeed
Warning

A335805-250MG A335805-1G A335805-5G
103825-702EA 40 USD
103825-702 103836-674 76803-082 77306-494 77306-496
2-(Pyrimidin-5-yl)benzaldehyde 95%
2-(Pyrimidin-5-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O
MW: 184.1900 g/mol
Boiling Pt: 375.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02684096
CAS Number: 640769-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR